Learn More
Olsalazine Sodium, Mikromol™
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
Description
Olsalazine Sodium
Specifications
Specifications
| Chemical Name or Material | Olsalazine sodium |
| CAS | 6054-98-4 |
| Analyte or Component Names | Olsalazine sodium |
| Shipping Condition | Room Temperature |
| Molecular Formula | C14 H8 N2 O6 . 2 Na |
| Certifications/Compliance | ISO 17034 |
| Synonym | Olsalazine sodium, Disodium 3,3'-diazenediylbis(6-hydroxybenzoate), Benzoic acid, 3,3'-(1,2-diazenediyl)bis[6-hydroxy-, sodium salt (1:2), Benzoic acid, 3,3'-azobis[6-hydroxy-, disodium salt (9CI), C.I. Mordant Yellow 5 (7CI), C.I. Mordant Yellow 5, disodium salt (8CI), 3,3'-Azobis[6-hydroxybenzoic acid] disodium salt, Acid Chrome Flavine R, Alizarol Flavine A, Azodisal sodium, C.I. 14130, Chromaven Yellow FA, Chrome Fast Flavine A, Chrome Fast Flavine J, Chrome Yellow A extra, Chrome Yellow AS, Curolite Yellow 3R, Diamond Chrome Yellow A ex, Dipentum, Disodium 3,3'-azobis(6-hydroxybenzoate), Disodium azodisalicylate, Eriochromal Flavine A, Eriochrome Flavine A, Fenakrom Yellow 3G, Metachrome Yellow A extra, Monochrome Yellow A ex, Mordant Yellow 5, Nutrachrome Yellow NYA, Olsalazine Disodium, Omega Chrome Aurine GL, Ph CJ 91B, Pontachrome Flavine A, Salicine Chrome Yellow A Extra, Sodium azodisalicylate, Solochrome Flavine G, Synchromate Flavine A, Telon Chrome Yellow T, Tertrochrome Yellow A |
| InChI Formula | InChI=1S/C14H10N2O6.2Na/c17-11-3-1-7(5-9(11)13(19)20)15-16-8-2-4-12(18)10(6-8)14(21)22;;/h1-6,17-18H,(H,19,20)(H,21,22);;/q;2*+1/p-2/b16-15+;; |
| SMILES | [Na+].[Na+].Oc1ccc(cc1C(=O)[O-])N=Nc2ccc(O)c(c2)C(=O)[O-] |
| IUPAC Name | disodium;5-[(E)-(3-carboxylato-4-hydroxyphenyl)diazenyl]-2-hydroxybenzoate |
| Show More |