missing translation for 'onlineSavingsMsg'
Learn More

Olsalazine Disodium, MedChemExpress

Product Code. p-200101493
Encompass_Preferred
Click to view available options
Quantity:
5 g
10 g
Unit Size:
10g
5g
This item is not returnable. View return policy

Product Code. 16470798

Brand: Medchem Express HYB0174

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Olsalazine Disodium is an anti-inflammatory drug used in the treatment of Inflammatory Bowel Disease and Ulcerative Colitis.

Olsalazine Disodium is an anti-inflammatory drug used in the treatment of Inflammatory Bowel Disease and Ulcerative Colitis.

Specifications

Chemical Name or Material Olsalazine Disodium
CAS 6054-98-4
Color Yellow
Purity Grade Notes Research
Molecular Formula C14H8N2Na2O6
Quantity 5 g
Solubility Information H2O : 50 mg/mL (144.43 mM; Need ultrasonic) ∣DMSO : 20 mg/mL (57.77 mM; Need ultrasonic)
SMILES O=C(O[Na])C1=C(O)C=CC(/N=N/C2=CC(C(O[Na])=O)=C(O)C=C2)=C1
Molecular Weight (g/mol) 346.2
Formula Weight 346.2
Percent Purity 99.36%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.