Learn More
Olsalazine Dimethyl Ester, TRC
Specifications
Specifications
| Chemical Name or Material | Olsalazine Dimethyl Ester |
| CAS | 101351-60-4 |
| Molecular Formula | C16H14N2O6 |
| Quantity | 500 mg |
| Synonym | 5,5'-Azodisalicyclic Dimethyl Ester |
| InChI Formula | InChI=1S/C16H14N2O6/c1-23-15(21)11-7-9(3-5-13(11)19)17-18-10-4-6-14(20)12(8-10)16(22)24-2/h3-8,19-20H,1-2H3 |
| SMILES | COC(=O)c1cc(ccc1O)N=Nc2ccc(O)c(c2)C(=O)OC |
| IUPAC Name | methyl 2-hydroxy-5-[(4-hydroxy-3-methoxycarbonylphenyl)diazenyl]benzoate |
| Molecular Weight (g/mol) | 330.29 |
| Formula Weight | 330.0852 |