Learn More
Olsalazine Dimethyl Ester, TRC
Product Code. 30416682
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30416682 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30416682 Supplier Toronto Research Chemicals Supplier No. TRCO586010100MG
 Restricted Item
Estimated Shipment: 12-11-2026
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Olsalazine Dimethyl Ester
CAS 101351-60-4
Molecular Formula C16H14N2O6
Quantity 100 mg
Synonym 5,5'-Azodisalicyclic Dimethyl Ester
InChI Formula InChI=1S/C16H14N2O6/c1-23-15(21)11-7-9(3-5-13(11)19)17-18-10-4-6-14(20)12(8-10)16(22)24-2/h3-8,19-20H,1-2H3
SMILES COC(=O)c1cc(ccc1O)N=Nc2ccc(O)c(c2)C(=O)OC
IUPAC Name methyl 2-hydroxy-5-[(4-hydroxy-3-methoxycarbonylphenyl)diazenyl]benzoate
Molecular Weight (g/mol) 330.29
Formula Weight 330.0852