Learn More
Olivetol-d9, TRC
Click to view available options
Quantity:
1 mg
10 mg
Unit Size:
1mg
Specifications
Specifications
| Chemical Name or Material | Olivetol-d9 |
| CAS | 137125-92-9 |
| Molecular Formula | C11H7D9O2 |
| Quantity | 1 mg |
| Synonym | 3,5-Dihydroxyamylbenzene-d9, 5-n-Amylresorcinol-d9, 5-n-Pentylresorcinol-d9, 1,3-Dihydroxy-5-pentylbenzene-d9, 5-Pentyl-1,3-benzenediol-d9; |
| SMILES | OC1=CC(O)=CC(CC([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])[2H])=C1 |
| Molecular Weight (g/mol) | 189.3 |
| Formula Weight | 189.1715 g/mol |
| Percent Purity | >95% (HPLC) |