Learn More
Oleic acid, 99%
Product Code. p-7049957
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
Product Code. Quantity unitSize
11398307 5 g 5g
11388307 25 g 25g
11410530 100 g 100g
3 options
Product Code. 11398307
Brand: Thermo Scientific Alfa Aesar 031997.06
Request Bulk or Custom Format
CAS: 112-80-1 | C18H34O2 | 282.47 g/mol
Oleic acid is used as an emulsifying agent for facial creams and lotions, as an accelerator in the rubber industry, as an emollient, as a solubilizing agent in aerosol products and as a soldering flux in stained glass work for joining lead came. It is used in the preparation of oleates and lotions. It acts as intermediate for drilling and fracking fluids in the oil and gas industry. It finds application as an excipient in pharmaceuticals.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 112-80-1
Molecular Formula C18H34O2
Molecular Weight (g/mol) 282.47
MDL Number MFCD00064242
InChI Key ZQPPMHVWECSIRJ-MDZDMXLPSA-N
Synonym oleic acid, cis-9-octadecenoic acid, cis-oleic acid, elaidoic acid, oleate, glycon wo, wecoline oo, pamolyn 100, glycon ro, z-octadec-9-enoic acid
PubChem CID 445639
ChEBI CHEBI:16196
IUPAC Name (Z)-octadec-9-enoic acid
SMILES CCCCCCCC\C=C\CCCCCCCC(O)=O
Specifications
Melting Point 13°C to 14°C
Color Colorless to Yellow
Density 0.887 g/mL
Boiling Point 286°C (100 mmHg)
Refractive Index 1.461
Quantity 5 g
Beilstein 1726542
Sensitivity Air Sensitive
Merck Index 146828
Solubility Information Miscible with ethanol,ether,acetone,chloroform,dimethyl formamide and dimethyl sulfoxide.
Formula Weight 282.47
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material Oleic acid