Learn More
Olanzapine Thiolactam Impurity, TRC
Product Code. 18023930
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
Unit Size:
10mg
1mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18023930 10 mg 10mg
18040920 5 mg 5mg
18030530 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 18023930 Supplier Toronto Research Chemicals Supplier No. TRCO25378010MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 1017241-36-9
Molecular Formula C17 H20 N4 O S
Molecular Weight (g/mol) 328.43
Synonym (1Z)-1-[4,5-Dihydro-2-(4-methyl-1-piperazinyl)-4-thioxo-3H-1,5-benzodiazepin-3-ylidene]-2-propanone, (Z)-Olanzapine Ketothiolactam
IUPAC Name (1Z)-1-[4-(4-methylpiperazin-1-yl)-2-sulfanylidene-1H-1,5-benzodiazepin-3-ylidene]propan-2-one
SMILES CN1CCN(CC1)C2=Nc3ccccc3NC(=S)/C/2=C\C(=O)C
Specifications
Quantity 10 mg
InChI Formula InChI=1S/C17H20N4OS/c1-12(22)11-13-16(21-9-7-20(2)8-10-21)18-14-5-3-4-6-15(14)19-17(13)23/h3-6,11H,7-10H2,1-2H3,(H,19,23)/b13-11-
Formula Weight 328.1358
Percent Purity >95%
Chemical Name or Material (Z)-Olanzapine Ketothiolactam
Grade HPLC