Learn More
Euro Protection™ Okenite Safety Shoes
Product Code. 12024295
Change view
Click to view available options
Size:
37
38
39
41
42
44
45
Unit Size:
Each
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size Unit Size
12024295 38 Each
12014295 37 Each
12034295 39 Each
12054295 41 Each
12064295 42 Each
12084295 44 Each
12094295 45 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Product Code. 12024295 Supplier Euro Protection™ Supplier No. 9OKEL38
  • ONLINE EXCLUSIVE PRICE
    792.75 NOK
    429.44 NOK / Pack of 1
    Save 363.31 NOK
    45% Off
Estimated Shipment: 08-10-2026
Euro Protection™ Okenite Safety Shoes are breathable, easy to put on/take off, suitable for food industry use, and feature a composite toe cap for protection.
  • Suitable for food industry use
  • Easy-on and easy-off
  • Protective cover
  • Rounded heel for more comfortable driving
  • Microfiber upper
  • Breathable 3D mesh lining
  • Removable, textile and foam insole
  • Composite toe cap
  • Injected polyurethane midsole
  • Polyurethane outsole
Specifications
Color White, Gray
For Use With (Application) Suitable for food industry
Heel Style Rounded
Liner Material Breathable 3D mesh lining
Material Microfiber
Size 38
Sole Features Removable, textile and foam insole, injected polyurethane midsole, polyurethane outsole.
Sole Material Polyurethane
Style Low-cut
Toe Material Composite
Weight (Metric) 427 g per shoe in size 8
Certifications/Compliance (Europe) EN ISO 20345 : 2011
Product Type Safety Shoe