Learn More
Ohaus™ OIML F2 Weight Set
Product Code. 16612212 Shop All Ohaus Products
Change view
Click to view available options
Mass:
1 kg to 1 g
1 kg to 1 mg
100 g to 1 g
100 g to 1 mg
2 x 2 kg to 1 g
2 x 2 kg to 1 mg
2 x 200 g to 1 g
2 x 200 g to 1 mg
5 kg to 1 g
5 kg to 1 mg
50 g to 1 g
50 g to 1 mg
500 g to 1 g
500 g to 1 mg
500 mg to 1 mg
Unit Size:
Each
Product Code. Mass unitSize
16612212 50 g to 1 mg Each
16622212 500 mg to 1 mg Each
16602212 50 g to 1 g Each
16692202 100 g to 1 mg Each
16682202 100 g to 1 g Each
16632192 2 x 200 g to 1 mg Each
16622192 2 x 200 g to 1 g Each
16672202 500 g to 1 mg Each
16662202 500 g to 1 g Each
16652202 1 kg to 1 mg Each
16642202 1 kg to 1 g Each
16612192 2 x 2 kg to 1 mg Each
16602192 2 x 2 kg to 1 g Each
16632202 5 kg to 1 mg Each
16622202 5 kg to 1 g Each
15 options
Product Code. 16612212
Brand: Ohaus™ 30390827
Only null left
Add to basket
Add to basket
Adjusted to Meet or Exceed OIML F2 Tolerances
  • Weights in masses ranging from 1 gram to 1 kilogram are made of Troemner Alloy 8 Stainless Steel which possesses closely controlled density (8.03 g/cm3), extremely low magnetic properties, good stability, and resistance to corrosion
  • Weights are supplied in high quality cases to ensure weights are held securely, kept clean, and dust free, protecting your investment
  • Weight surfaces meet or exceed OIML R 111 and have a highly polished mirror-like finish which allows scratches and foreign matters on the weight to be easily seen allowing for stability and longevity
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance No Certificate
Material Stainless Steel, Aluminum
Mass 50 g to 1 mg
Includes Protective Case
Packaging Quantity 20/Pk.