Learn More
Sartorius™ OIML Class F1 Proof Line Knob Weight without Certificate
Product Code. 11373304 Shop All Sartorius Products
Click to view available options
Mass:
1 g
2 g
5 g
10 g
20 g
50 g
100 g
200 g
500 g
1 kg
2 kg
5 kg
10 kg
Unit Size:
Each
Product Code. 11373304
Brand: Sartorius™ YCW613AC00
Only null left
Add to basket
Add to basket
Polished non-magnetizable calibration weights. Sartorius™ OIML Class F1 Knob Weight without Certificate is constructed of special steel and supplied in a plastic case.
  • Non-magnetizable
  • Supplied in a high-quality plastic case (unless otherwise specified) suitable for use in cleanrooms
  • Special Stainless steel construction with a density of 7,950 kg/m3
  • Complies with OIML R111-1 International Recommentation
TRUSTED_SUSTAINABILITY
Specifications
Product Type Proof Line Weight
Classification OIML Class F1
Material Special Stainless Steel
Density 7,950kg/m3
Mass 1 kg