Learn More
Kern Sohn OIML Class F1 Knob Test Weight
Product Code. 17392093
Encompass_Preferred
Change view
Click to view available options
Weight (Metric):
1 g
2 g
5 g
10 g
20 g
50 g
100 g
200 g
1 kg
2 kg
5 kg
10 kg
20 kg
50 kg
500 kg
Unit Size:
Each
Product Code. Weight (Metric) unitSize
17392093 10 kg Each
17372083 1 g Each
17382083 2 g Each
17392083 5 g Each
17302093 10 g Each
17312093 20 g Each
17322093 50 g Each
17332093 100 g Each
17342093 200 g Each
17352093 500 kg Each
17362093 1 kg Each
17372093 2 kg Each
17382093 5 kg Each
17302103 20 kg Each
17312103 50 kg Each
15 options
Product Code. 17392093
Brand: Kern Sohn 32714
Only null left
Add to basket
Add to basket
Single test weight constructed of stainless steel.
  • Knob shape
  • Polished stainless steel
Specifications
Classification F1
Material Stainless Steel
Weight (Metric) 10 kg
Tolerance ± 50 mg