Learn More
Kern Sohn OIML Class F1 ECO-Shape Test Weight
Product Code. p-7226244
Encompass_Preferred
Change view
Click to view available options
Weight (Metric):
1 g
2 g
5 g
10 g
20 g
50 g
100 g
200 g
500 g
1 kg
2 kg
5 kg
Unit Size:
Each
Product Code. Weight (Metric) unitSize
17302073 1 g Each
17312073 2 g Each
17322073 5 g Each
17332073 10 g Each
17342073 20 g Each
17352073 50 g Each
17362073 100 g Each
17372073 200 g Each
17382073 500 g Each
17392073 1 kg Each
17302083 2 kg Each
17312083 5 kg Each
12 options
Product Code. 17302073
Brand: Kern Sohn 32601A
This item requires a price update prior to being ordered.
Request a Price for this item here.
Single test weight constructed of stainless steel.
  • ECO-shape
  • Polished stainless steel
Specifications
Classification F1
Material Stainless Steel
Weight (Metric) 1 g
Tolerance ± 0.1 mg