Learn More
Kern Sohn OIML Class F1 ECO-Shape Test Weight
Product Code. 17392073
Encompass_Preferred
Click to view available options
Weight (Metric):
1 g
2 g
5 g
10 g
20 g
50 g
100 g
200 g
500 g
1 kg
2 kg
5 kg
Unit Size:
Each
Product Code. 17392073
Brand: Kern Sohn 32611A
Only null left
Add to basket
Add to basket
Single test weight constructed of stainless steel.
  • ECO-shape
  • Polished stainless steel
Specifications
Classification F1
Material Stainless Steel
Weight (Metric) 1 kg
Tolerance ± 5 mg
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.