Learn More
Kern Sohn OIML Class F1 ECO-Shape Test Weight
Description
- ECO-shape
- Polished stainless steel
Specifications
Specifications
| Classification | F1 |
| Material | Stainless Steel |
| Weight (Metric) | 200 g |
| Tolerance | ± 1 mg |