Learn More
3M™ Oil Sorbent Boom
Product Code. 17118650
Change view
Click to view available options
Diameter (Metric):
100 mm
200 mm
Length (Metric):
3 m
5 m
Unit Size:
Pack of 2
Pack of 4
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Diameter (Metric) Length (Metric) unitSize
17118650 100 mm 3 m Pack of 4
17128650 200 mm 3 m Pack of 4
17108650 200 mm 5 m Pack of 2
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 17118650 Supplier 3M™ Supplier No. 7000088228
  • ONLINE EXCLUSIVE PRICE
    7835.00 NOK
    6349.81 NOK / Pack of 4
    Save 1485.19 NOK
    18% Off
Estimated Shipment: 29-09-2026
Add to basket
Add to basket
A double boom with two, 4 inch scrims encased tubes of particulate sorbent material, secured by a UV resistant netting.
  • Used to cover large spills, or as a precaution in areas where large spills may occur
  • Ideal for absorbing oil from areas where water may also be present
  • Link sorbent booms together by built-in connectors to deploy over large areas
  • UV-resistant to prevent deterioration and disintegration
  • Designed for long-term use in ditches, streams, rivers, lakes, docks, and piers
  • Cost-effective as oil, not water, is recovered
  • Can be used outside or in-plant
TRUSTED_SUSTAINABILITY
Specifications
Product Type Sorbent Boom
Color Brown
Diameter (Metric) 100 mm
For Use With (Application) Workplace Safety, Spill Prevention
Length (Metric) 3 m
Model T-280