Learn More
Draeger Safety Oil Impactor
Product Code. 15644994
Change view
Click to view available options
Quantity:
10/Pk.
Unit Size:
Pack of 10
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15644994 10/Pk. Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15644994 Supplier Draeger Safety Supplier No. 8103530
  • ONLINE EXCLUSIVE PRICE
    3274.88 NOK
    1912.59 NOK / Pack of 10
    Save 1362.29 NOK
    41% Off
Estimated Shipment: 18-12-2026
Draeger Safety Oil Impactor tests for all synthetic and mineral oil mists in compressed-air and breathing-air.
  • Utilizing an innovative oil detection process the impactor eliminates the need to use an oxidizing reaction to detect for the presence of oil
Specifications
Description Wide
Quantity 10/Pk.