Learn More
Thermo Scientific™ Oil-Free Vacuum Pumps for Vacuum Concentrators
Product Code. p-4528998 Shop All Thermo Scientific Products
Change view
Click to view available options
Description:
EFTE/PFA 4-headed Vacuum Pump
Voltage:
230 V
Unit Size:
Each
Product Code. Description Voltage unitSize
10543234 EFTE/PFA 4-headed Vacuum Pump 230 V Each
1 options
Product Code. 10543234
Brand: Thermo Scientific™ OFP400230
Only null left
Add to basket
Add to basket
Eliminate oil mist vapors with these quiet-running, chemical resistant, oil-free pumps, designed for a wide variety of laboratory applications.
Eliminate oil mist vapors with Thermo Scientific™ Oil-Free Vacuum Pumps for Vacuum Concentrators, designed for a wide variety of laboratory uses. These quiet-running pumps are highly chemical resistant, and do not require oil to operate.
  • Small footprint
  • Four-headed design with chemically resistant solid ETFE and PFA heads
  • Easily connects to any Thermo Scientific™ SpeedVac™ system
  • Recommended for use with acidic environments such as HCl, TFA, Acetic and Formic Acids as well as organic solvents
  • Comes complete with ANT100 Post Trap for adding trapping and neutralizing solutions
TRUSTED_SUSTAINABILITY
Specifications
Description EFTE/PFA 4-headed Vacuum Pump
Type Oil-Free Vacuum Pump
Displacement 36L/min. at 60Hz
For Use With (Equipment) Vacuum Concentrator
Max. Vacuum 0.6 torr (0.75 mbar)
Dimensions (L x W x H) 15 x 8 x 18 in. (390 x 210 x 460 mm)
Length (English) 15 in.
Length (Metric) 390 mm
Width (English) 8 in.
Width (Metric) 210 mm
Height (English) 18 in.
Height (Metric) 460 mm
Shipping Weight (English) 49.5 lb.
Shipping Weight (Metric) 22.5 kg
Electrical Requirements 230 V 50/60 Hz
Voltage 230 V
Frequency 50/60 Hz