Description
Counterweight set for use with the following mechanical scales:
- Triple Beam 710-00
- Triple Beam 750-S0
- Triple Beam 760-00
- Triple Beam Dial-O-Gram 1650-00
- Triple Beam Pro TP2611
Specifications
Specifications
| Product Type | Metric Attachment Weight Set |
| Description | Weight Set, 2x1kg, 1x500g |
| Classification | Miscellaneous |
| Material | Cast Iron |
| Weight (Metric) | 1 x 500 g, 2 x 1 kg |
| Shape | Cylindrical |
| For Use With (Application) | Calibration, counter weight |
| For Use With (Equipment) | Triple Beam 710-00, 750-S0 and 760-00; Triple Beam Dial-O-Gram 1650-00; and Triple Beam Pro TP2611 |
| Packaging Quantity | 3 Weights/Pk. |