missing translation for 'onlineSavingsMsg'
Learn More
Learn More
OGG1 Antibody (4E8) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-52722
This item is not returnable.
View return policy
Description
OGG1 Monoclonal specifically detects OGG1 in Human samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Flow (Intracellular).
Specifications
| OGG1 | |
| Monoclonal | |
| 1.0 mg/mL | |
| Flow Cytometry 2-5 ug/ml, Immunohistochemistry 5 ug/ml, Immunocytochemistry/Immunofluorescence 10 ug/ml, Immunohistochemistry-Paraffin 5 ug/ml, Flow (Intracellular) | |
| 8-hydroxyguanine DNA glycosylase, 8-oxoguanine DNA glycosylase, AP lyase, DNA-apurinic or apyrimidinic site lyase, HOGG1, MMH, MUTMOGH1HMMH, N-glycosylase/DNA lyase | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| 4968 | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG2b |
| Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| 400000000 | |
| Unconjugated | |
| PBS with 0.05% Sodium Azide | |
| OGG1 | |
| Partial recombinant protein made to an internal sequence of the human OGG1 protein (between amino acids 100-300) [UniProt# O15527] | |
| 0.1 mg | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction