Learn More
OGDH Mouse anti-Human, Unconjugated, Clone: 1H7A2, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 66285-1-IG-20UL
Additional Details : Weight : 0.00010kg
Description
The OGDH antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human OGDH. This antibody recognizes human, rat antigen. The OGDH antibody has been validated for the following applications: IF, IHC, WB, ELISA analysis.
Specifications
OGDH | |
Monoclonal | |
1000 μg/mL | |
Mouse | |
Primary | |
Human, Rat |
Western Blot, Immunohistochemistry, Immunofluorescence, ELISA | |
1H7A2 | |
Unconjugated | |
20 μL | |
4967 | |
IgG2b |