Learn More
OGDH Mouse anti-Human, Unconjugated, Clone: 1H7A2, Proteintech
Mouse Monoclonal Antibody
Brand:  Proteintech 66285-1-IG-150UL
Additional Details : Weight : 0.00010kg
 View more versions of this product
Product Code. 16888015
  • 4593.76 NOK / 150µL
Estimated Shipment: 22-05-2024
Description
The OGDH antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human OGDH. This antibody recognizes human, rat antigen. The OGDH antibody has been validated for the following applications: IF, IHC, WB, ELISA analysis.
Specifications
Specifications
OGDH
Monoclonal
1000 μg/mL
Mouse
Primary
Human, Rat
Western Blot, Immunohistochemistry, Immunofluorescence, ELISA
1H7A2
Unconjugated
150 μL
4967
IgG2b