Description
The OGDH antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human OGDH. This antibody recognizes human, rat antigen. The OGDH antibody has been validated for the following applications: IF analysis. This antibody is coralite™594.
Specifications
Specifications
| Antigen | OGDH |
| Applications | Immunofluorescence |
| Classification | Monoclonal |
| Clone | 1H7A2 |
| Concentration | 2000 μg/mL |
| Conjugate | CoraLite 594 |
| Host Species | Mouse |
| Quantity | 100 μL |
| Primary or Secondary | Primary |
| Gene ID (Entrez) | 4967 |
| Show More |