Learn More
OFD1 Rabbit anti-Human, Polyclonal, Proteintech
Product Code. 16802115 Shop All Proteintech Products
Change view
Click to view available options
Quantity:
150 μL
20 μL
Unit Size:
150µL
20µL
Product Code. Quantity unitSize
16802115 20 μL 20µL
16892105 150 μL 150µL
2 options
Product Code. 16802115
Brand: Proteintech 228511AP20UL
Rabbit Polyclonal Antibody
This antibody recognize all the isoforms of OFD1. The 43 kDa is isoform 2. The ∽80 kDa band is unknown.
This gene is located on the X chromosome and encodes a centrosomal protein. A knockout mouse model has been used to study the effect of mutations in this gene. The mouse gene is also located on the X chromosome, however, unlike the human gene it is not subject to X inactivation. Mutations in this gene are associated with oral-facial-digital syndrome type I and Simpson-Golabi-Behmel syndrome type 2. Many pseudogenes have been identified; a single pseudogene is found on chromosome 5 while as many as fifteen have been found on the Y chromosome.
Specifications
Antigen OFD1
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.29 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene OFD1
Gene Accession No. O75665
Gene Alias 71 7A, CXorf5, OFD1, oral facial digital syndrome 1, Protein 71 7A, SGBS2
Gene Symbols OFD1
Host Species Rabbit
Immunogen OFD1 Fusion Protein Ag18898
Purification Method Antigen Affinity Chromatography
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8481
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG