missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ODC1 Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
1611.84 NOK - 4593.76 NOK
Specifications
Antigen | ODC1 |
---|---|
Concentration | 233 μg/mL |
Applications | Western Blot, Immunohistochemistry |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16886225
|
Proteintech
28728-1-AP-20UL |
20 μL |
1611.84 NOK
20µL |
Estimated Shipment: 15-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16876225
|
Proteintech
28728-1-AP-150UL |
150 μL |
4593.76 NOK
150µL |
Estimated Shipment: 15-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
The ODC1 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human ODC1. This antibody recognizes human, mouse antigen. The ODC1 antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
ODC1 | |
Western Blot, Immunohistochemistry | |
Unconjugated | |
Human, Mouse | |
IgG |
233 μg/mL | |
Polyclonal | |
Rabbit | |
4953 | |
Primary |