Learn More
Octylamine, Sigma-Aldrich™
Specifications
Specifications
| CAS | 111-86-4 |
| Melting Point | -1°C |
| Density | 0.78 g/cm3 at 20°C |
| Boiling Point | 178°C to 179°C/1013 hPa |
| Molecular Formula | CH3(CH2)7NH2 |
| MDL Number | MFCD00008247 |
| Quantity | 250 mL |
| Synonym | Octylamine, 1-Aminooctane, Caprylamine, Octylamine |
| Molecular Weight (g/mol) | 129.24 |
| Grade | Research |