Learn More
Octyl Sulfate Sodium Salt, Sigma-Aldrich™
Product Code. 15887900
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15887900 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
MQ200 Graded - For Research and Non-regulated Industrial Applications
Product Code. 15887900 Supplier Sigma Aldrich Supplier No. 1122680005
 Restricted Item
Estimated Shipment: 18-11-2026
MQ200 Graded - For Research and Non-regulated Industrial Applications
For surfactant tests
TRUSTED_SUSTAINABILITY
Specifications
CAS 142-31-4
Melting Point 195°C (Decomposition)
Concentration 100%
Quantity 5 g
MDL Number MFCD00007470
Synonym Octyl sulfate sodium salt, Sodium octyl sulfate, Octyl sulfate sodium salt
Molecular Weight (g/mol) 232.27
Chemical Name or Material Octyl Sulfate Sodium Salt