Learn More
MedChemExpress Octreotide
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
Specifications
Specifications
| Purity | 98.84% |
| CAS | 83150-76-9 |
| Content And Storage | Protect from light∣Powder -80°C 2 years, -20°C 1 year∣The compound is unstable in solutions, freshly prepared is recommended. |
| For Use With (Application) | COVID-19-immunoregulation |
| Molecular Weight (g/mol) | 1019.24 g/mol |
| Product Type | Peptides |
| Quantity | 25 mg |
| Shipping Condition | Room temperature |
| Solubility | H2O : 100 mg/mL (98.11 mM∣Need ultrasonic) |
| Synonym | SMS 201-995 |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.