Learn More
Octanoic Acid, Sigma-Aldrich™
Product Code. 15838190
Change view
Click to view available options
Quantity:
100 mL
1 L
Unit Size:
1 L
100mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15838190 1 L 1 L
15828190 100 mL 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15838190 Supplier Sigma Aldrich Supplier No. 8001921000
 Restricted Item
Estimated Shipment: 26-11-2026
For synthesis
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 124-07-2
Specifications
Melting Point 16°C
Density 0.91 g/cm3 at 20°C
Boiling Point 237°C/1013 hPa
Molecular Formula CH3(CH2)6COOH
MDL Number MFCD00004429
Quantity 1 L
Synonym Octanoic acid, Octanoic acid
Molecular Weight (g/mol) 144.21
Grade Research