Learn More
Octamethylcyclotetrasiloxane, Sigma-Aldrich™
Product Code. 15884351
Change view
Click to view available options
Quantity:
25 mL
100 mL
250 mL
Unit Size:
100mL
250mL
25mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15884351 250 mL 250mL
15864351 25 mL 25mL
15874351 100 mL 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 15884351 Supplier Sigma Aldrich Supplier No. 8147500250
 Restricted Item
Estimated Shipment: 18-11-2026
For synthesis
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 556-67-2
Specifications
Melting Point 18°C
Density 0.95 g/cm3 at 25°C
Boiling Point 176°C/1013 hPa
MDL Number MFCD00003269
Quantity 250 mL
Synonym Octamethylcyclotetrasiloxane, Octamethyltetrasiloxane, Octamethylcyclotetrasiloxane
Molecular Weight (g/mol) 296.62
Grade Research