Learn More
Octahydrocyclopenta[b]morpholine, TRC
Specifications
Specifications
| Chemical Name or Material | Octahydrocyclopenta[b]morpholine |
| CAS | 1018639-83-2 |
| Molecular Formula | C7 H13 N O |
| Quantity | 10 mg |
| Synonym | Octahydrocyclopent[b]-1,4-oxazine, 2,3,4,4a,5,6,7,7a-Octahydrocyclopenta[b][1,4]oxazine, Octahydrocyclopenta[b][1,4]oxazine |
| InChI Formula | InChI=1S/C7H13NO/c1-2-6-7(3-1)9-5-4-8-6/h6-8H,1-5H2 |
| SMILES | C1CC2NCCOC2C1 |
| IUPAC Name | 2,3,4,4a,5,6,7,7a-octahydrocyclopenta[b][1,4]oxazine |
| Molecular Weight (g/mol) | 127.184 |
| Formula Weight | 127.1 |