Learn More
Octadecylamine, Sigma-Aldrich™
Specifications
Specifications
| CAS | 124-30-1 |
| Melting Point | 50°C to 52°C |
| Density | 0.86 g/cm3 at 20°C |
| Boiling Point | 132°C/43 hPa |
| MDL Number | MFCD00008159 |
| Quantity | 250 g |
| Synonym | Octadecylamine, 1-Aminooctadecane, Stearylamine, Octadecylamine |
| Molecular Weight (g/mol) | 269.51 |
| Grade | Research |