Learn More
- Home
- Products
- Chromatography Columns and Cartridges
- Column Packing
- Macherey-Nagel™ Octadecyl Phase Polygosil™ C18
Macherey-Nagel™ Octadecyl Phase Polygosil™ C18
Product Code. 10746692 Shop All Macherey-Nagel Products
Change view
Click to view available options
Quantity:
10 g
100 g
Particle Size:
5 μm
7 μm
10 μm
Pore Size:
100 Å
1000 Å
300 Å
60 Å
Unit Size:
100g
10g
| Product Code. | Quantity | Particle Size | Pore Size | unitSize |
|---|---|---|---|---|
| 10746692 | 10 g | 5 μm | 60 Å | 10g |
| 10778222 | 100 g | 10 μm | 100 Å | 100g |
| 10104601 | 100 g | 10 μm | 60 Å | 100g |
| 10505961 | 100 g | 5 μm | 100 Å | 100g |
| 10641292 | 100 g | 5 μm | 60 Å | 100g |
| 10515781 | 100 g | 7 μm | 1000 Å | 100g |
| 10566121 | 100 g | 7 μm | 300 Å | 100g |
| 10194591 | 100 g | 7 μm | 60 Å | 100g |
| 10022681 | 10 g | 10 μm | 100 Å | 10g |
| 10419351 | 10 g | 10 μm | 60 Å | 10g |
| 10595951 | 10 g | 5 μm | 100 Å | 10g |
| 10296401 | 10 g | 7 μm | 1000 Å | 10g |
| 10032741 | 10 g | 7 μm | 300 Å | 10g |
| 10595771 | 10 g | 7 μm | 60 Å | 10g |
This item is not returnable.
View return policy
This item is not returnable.
View return policy
Specifications
Specifications
| Particle Size | 5 μm |
| Pore Size | 60 Å |
| Technique | Liquid Chromatography |
| Quantity | 10 g |
| Type | Polygosil C18 Phases |