Learn More
OCT1 Rabbit anti-Human, Porcine, Polyclonal, Proteintech
Product Code. 16846743 Shop All Proteintech Products
Change view
Click to view available options
Quantity:
150 μL
20 μL
Unit Size:
150µL
20µL
Product Code. Quantity unitSize
16846743 20 μL 20µL
16836743 150 μL 150µL
2 options
Product Code. 16846743
Brand: Proteintech 103871AP20UL
Rabbit Polyclonal Antibody
The OCT1 transcription factor was among the first identified members of the POU transcription factor family. Members of this family contain the POU domain, a 160-amino acid region necessary for DNA binding to the octameric sequence ATGCAAAT.
Specifications
Antigen OCT1
Applications Immunoprecipitation, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.23 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Gene POU2F1
Gene Accession No. P14859, Q29076
Gene Alias NF A1, Oct 1, OCT1, Octamer binding protein 1, OTF 1, OTF1, POU class 2 homeobox 1, POU2F1
Gene Symbols POU2F1
Host Species Rabbit
Immunogen OCT1 Fusion Protein Ag0586
Purification Method Antigen Affinity Chromatography
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 397501, 5451
Target Species Pig, Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG