Learn More
OCIAD1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 16634-1-AP-150UL
 View more versions of this product
Product Code. 16846314
  • 4836.10 NOK / 150µL
Description
This antibody detects OCIAD1 with an apparent molecular weight of ∽35 kDa as has been demonstrated by several researches.
The Ras-related superfamily of guanine nucleotide binding proteins, which includes the R-Ras, Rap, Ral/Rec and Rho/Rab subfamilies, exhibit 30-60% homology with Ras p21. Accumulating data suggests an important role for Rab proteins, either in endocytosis or in biosynthetic protein transport. At least eight members of the Rab subfamily have been identified, each of which is found at a particular stage of a membrane transport pathway.
Specifications
Specifications
OCIAD1
Polyclonal
Unconjugated
OCIAD1
Asrij, OCIA, OCIA domain containing 1, OCIAD1, TPA018
Rabbit
Antigen Affinity Chromatography
RUO
289590, 54940, 68095
-20°C
Liquid
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
0.31 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q5XIG4, Q9CRD0, Q9NX40
Ociad1
OCIAD1 Fusion Protein Ag9977
150 μL
Primary
Human, Mouse, Rat