Learn More
Occludin Mouse anti-Human, Mouse, Porcine, Rat, Clone: 1D3C4, Proteintech
Mouse Monoclonal Antibody
1710.33 NOK - 4836.10 NOK
Tekniske data
| Antigen | Occludin |
|---|---|
| Clone | 1D3C4 |
| Concentration | 1 mg/mL |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Monoclonal |
| Produktkode | Brand | Quantity | Pris | Antal & tilgængelighed | |||||
|---|---|---|---|---|---|---|---|---|---|
| Produktkode | Brand | Quantity | Pris | Antal & tilgængelighed | |||||
|
16818205
|
Proteintech
66378-1-IG-20UL |
20 μL |
1710.33 NOK
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
16808205
|
Proteintech
66378-1-IG-150UL |
150 μL |
4836.10 NOK
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Beskrivelse
This gene encodes an integral membrane protein that is required for cytokine-induced regulation of the tight junction paracellular permeability barrier. Mutations in this gene are thought to be a cause of band-like calcification with simplified gyration and polymicrogyria (BLC-PMG), an autosomal recessive neurologic disorder that is also known as pseudo-TORCH syndrome. Alternative splicing results in multiple transcript variants. A related pseudogene is present 1.5 Mb downstream on the q arm of chromosome 5.Tekniske data
| Occludin | |
| 1 mg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Occludin, OCLN | |
| Ocln | |
| IgG1 | |
| Protein G | |
| Antibody |
| 1D3C4 | |
| Immunohistochemistry (Paraffin), Western Blot | |
| Unconjugated | |
| Mouse | |
| Mouse, Human, Pig, Rat | |
| Q16625, Q61146, Q6P6T5 | |
| 100506658, 18260, 397236, 83497 | |
| Occludin Fusion Protein Ag4057 | |
| Primary | |
| -20°C | |
| OCLN |
Ser du en mulighed for forbedring?Del en indholdskorrektion
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel