Learn More
Motic™ Objectives, Stereo for SMZ-161/171
Click to view available options
Magnification Power:
0.3X
0.63X
0.75X
1.5X
Description:
WD: 128.6mm
WD: 142.7mm
WD: 301mm
WD: 56.3mm
Unit Size:
Each
Description
- Suitable for SMZ-161 and SMZ-171 Microscopes.
Specifications
Specifications
| Type | Stereo |
| Description | WD: 56.3mm |
| For Use With (Equipment) | SMZ-161/171 Microscopes |
| Magnification Power | 1.5X |