Learn More
Motic™ Objective, Plan, Strain-free, CCIS EC for BA310
Color-corrected infinity system, enhanced-contrast Plan objective. Motic™ Objective, Plan, Strain-free, CCIS EC for BA310 is designed for field curvature correction and strain-free use.
Brand:  Motic™ 1101001703971
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 15465272
  • 4115.00 NOK / Each
Estimated Shipment: 26-07-2024
to see stock.
Add to basket
Description
Description
  • Suitable for BA310 Polarizing Microscope.
  • Color-corrected infinity system.
  • Enhanced contrast.
  • Field curvature correction.
  • Strain-free.
Specifications
Specifications
CCIS, EC Plan, Strain-free
NA: 0.65/S, WD: 0.5mm
BA310 Polarizing Microscope
40X