Learn More
Motic™ Objective, CCIS Plan Achromatic NCG EC PL for BA310E
Color-corrected infinity system, enhanced-contrast, non-cover glass and lead-free objective. Motic™ Objective, CCIS Plan Achromatic NCG EC PL for BA310E is designed for field curvature, spherical aberration, and chromatic aberration correction.
Brand:  Motic™ 1101001703001
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 15455242
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Description
Description
  • Suitable for BA310 Series Upright Compound Microscope.
  • Color-corrected infinity system.
  • Non-cover glass system.
  • Lead-free glass.
  • Enhanced contrast.
  • Field curvature correction.
  • Chromatic aberration correction for 2 colors.
  • Spherical correction for 1 color.
Specifications
Specifications
Objective
NA: 0.45, WD: 1mm
BA310E Microscopes
20X