Learn More
Motic™ Objective, CCIS Plan Achromatic NCG EC PL for BA310E
Color-corrected infinity system, enhanced-contrast, non-cover glass and lead-free objective. Motic™ Objective, CCIS Plan Achromatic NCG EC PL for BA310E is designed for field curvature, spherical aberration, and chromatic aberration correction.
Brand: Motic™ 1101001703001
Additional Details : Weight : 0.01000kg
Description
- Suitable for BA310 Series Upright Compound Microscope.
- Color-corrected infinity system.
- Non-cover glass system.
- Lead-free glass.
- Enhanced contrast. Field curvature correction.
- Chromatic aberration correction for 2 colors.
- Spherical correction for 1 color.
Specifications
Objective | |
NA: 0.45, WD: 1mm | |
BA310E Microscopes | |
20X |