Learn More
Motic™ Objective CCIS Plan Achromatic EC PL for BA310E
Click to view available options
Magnification Power:
100X
10X
20X
Description:
NA: 0.25, WD: 17.4mm
NA: 0.45mm, WD: 0.9mm
NA: 1.25/S-Oil, WD: 0.15mm
Unit Size:
Each
Description
- Suitable for BA310 Series Upright Compound Microscope.
- Color-corrected infinity system.
- Lead-free glass.
- Enhanced contrast. Field curvature correction.
- Chromatic aberration correction for 2 colors.
- Spherical correction for 1 color.
Specifications
Specifications
| Type | Objective |
| Description | NA: 0.45mm, WD: 0.9mm |
| For Use With (Equipment) | BA310E Microscopes |
| Magnification Power | 20X |