Learn More
Motic™ Objective CCIS Plan Achromatic EC PL for BA310E
Product Code. 15415232
Change view
Click to view available options
Magnification Power:
100X
10X
20X
Description:
NA: 0.25, WD: 17.4mm
NA: 0.45mm, WD: 0.9mm
NA: 1.25/S-Oil, WD: 0.15mm
Unit Size:
Each
Product Code. Magnification Power Description unitSize
15415232 10X NA: 0.25, WD: 17.4mm Each
15425232 20X NA: 0.45mm, WD: 0.9mm Each
15445232 100X NA: 1.25/S-Oil, WD: 0.15mm Each
3 options
Product Code. 15415232
Brand: Motic™ 1101001703221
Only null left
Add to basket
Add to basket
Color-corrected infinity system, enhanced-contrast and lead-free objective. Motic™ Objective CCIS Plan Achromatic EC PL for BA310E is designed for field curvature, spherical aberration, and chromatic aberration correction.
  • Suitable for BA310 Series Upright Compound Microscope.
  • Color-corrected infinity system.
  • Lead-free glass.
  • Enhanced contrast.
  • Field curvature correction.
  • Chromatic aberration correction for 2 colors.
  • Spherical correction for 1 color.
Specifications
Type Objective
Description NA: 0.25, WD: 17.4mm
For Use With (Equipment) BA310E Microscopes
Magnification Power 10X