Learn More
Motic™ Objective, Achromatic EA 10X/0.25
Product Code. 15445102
Click to view available options
Magnification Power:
10X
Unit Size:
Each
Product Code. 15445102
Brand: Motic™ 12180162
Only null left
Add to basket
Add to basket
Designed with chromatic aberration correction for 2 colors. Motic™ Objective, Achromatic EA 10X/0.25 is spherically corrected for one color.
  • Achromatic.
  • Chromatic aberration correction for 2 colors.
  • Spherical correction for 1 color.
Specifications
Type Achromat Objective
Description NA: 0.25
Focus Distance 5 mm WD
For Use With (Equipment) PM Series, DM1802-A, SERIES 18, Series 28 Microscopes
Magnification Power 10X