Learn More
Motic™ Objective, Achromatic 40X/0.65/S
Click to view available options
Magnification Power:
40X
Unit Size:
Each
Description
- Achromatic.
- Chromatic aberration correction for 2 colors.
- Spherical correction for 1 color.
Specifications
Specifications
| Type | Achromat Objective |
| Description | NA: 0.65/S |
| For Use With (Equipment) | SFC-100, DM-111, F-1110 Microscopes |
| Magnification Power | 40X |