Learn More
Motic™ Objective, Achromatic 10X/0.25
Product Code. 15495082
missing translation for 'changeView'
missing translation for 'orderingAttributeHoverText'
Magnification Power:
10X
missing translation for 'unitSize':
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Magnification Power missing translation for 'unitSize'
15495082 10X Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 missing translation for 'options'
Product Code. 15495082 missing translation for 'mfr' Motic™ missing translation for 'supplierNo' 1101001701133
  • ONLINE EXCLUSIVE PRICE
    344.00 NOK
    300.54 NOK / Pack of 1
    Save 43.46 NOK
    12% Off
 Restricted Item
Estimated Shipment: 02-12-2026
Add to basket
Add to basket
Designed with chromatic aberration correction for 2 colors. Motic™ Objective, Achromatic 10X/0.25 is spherically corrected for one color.
  • Achromatic.
  • Chromatic aberration correction for 2 colors.
  • Spherical correction for 1 color.
Specifications
Type Achromat Objective
Description NA: 0.25
For Use With (Equipment) SFC-100, DM-111, F-1110 Microscopes
Magnification Power 10X