Learn More
Abnova™ OBFC1 Recombinant Protein
Brand:  Abnova™ H00079991-P01.25ug
Additional Details : Weight : 0.00010kg
 View more versions of this product
Product Code. 16134703
  • 6245.00 NOK / 25µg
Estimated Shipment: 27-06-2024
Description
ELISA, Western Blotting (Recombinant Protein), Antibody Production, Protein Array
Specifications
Specifications
AAH17400
50mM Tris HCl, 10mM reduced Glutathione, pH 8 in the Elution Buffer
66.6
8
Glutathione Sepharose 4 Fast Flow
25 ug
MQPGSSRCEEETPSLLWGLDPVFLAFAKLYIRDILDMKESRQVPGVFLYNGHPIKQVDVLGTVIGVRERDAFYSYGVDDSTGVINCICWKKLNTESVSAAPSAARELSLTSQLKKLQETIEQKTKIEIGDTIRVRGSIRTYREEREIHATAYYKVDDPVWNIQIARMLELPTIYRKVYDQPFHSSALEKEEALSNPGALDLPSLTSLLSEKAKEFLMENRVQSFYQQELEMVESLLSLANQPVIHSACSDQVNFKKDTTSKAIHSIFKNAIQLLQEKGLVFQKDDGFDNLYYVTREDKDLHRKIHRIIQQDCQKPNHMEKGCHFLHILACARLSIRPGLSEAVLQQVLELLEDQSDIVSTMEHYYTAF
AAF-44/FLJ22559/RP11-541N10.2/RPA-32/bA541N10.2
OBFC1
Wheat Germ (in vitro)
GST
Antibody Production, Protein Array, ELISA, Western Blot
79991
OBFC1 (Human) Recombinant Protein (P01)
In vitro wheat germ expression system
12.5% SDS-PAGE Stained with Coomassie Blue.
Wheat Germ (in vitro)
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
OBFC1
Human
Recombinant
Solution