Learn More
Obatoclax Mesylate, MedChemExpress
Product Code. 16487037
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
Product Code. 16487037
Brand: Medchem Express HY10969
MedChemExpress Obatoclax Mesylate (GX15-070 Mesylate), a BH3 mimetic, is a pan-BCL-2 family proteins inhibitor with a Ki of 220 nM for BCL-2. Obatoclax Mesylate induces autophagy-dependent cell death and targets cyclin D1 for proteasomal degradation. Obatoclax Mesylate has anti-cancer and broad-spectrum antiparasitic activity.
Obatoclax Mesylate (GX15-070 Mesylate), a BH3 mimetic, is a pan-BCL-2 family proteins inhibitor with a Ki of 220 nM for BCL-2. Obatoclax Mesylate induces autophagy-dependent cell death and targets cyclin D1 for proteasomal degradation. Obatoclax Mesylate has anti-cancer and broad-spectrum antiparasitic activity.
Specifications
Chemical Name or Material Obatoclax Mesylate
Physical Form Solid
CAS 803712-79-0
Color Black
Quantity 5 mg
Purity Grade Notes Research
Molecular Formula C21H23N3O4S
For Use With (Application) Cancer-Kinase/protease
Synonym GX15-070 Mesylate
Solubility Information DMSO : 12.5 mg/mL (30.23 mM; Need ultrasonic)
SMILES COC1=CC(C2=CC3=C(C=CC=C3)N2)=N/C1=C\C4=C(C=C(N4)C)C.CS(=O)(O)=O
Molecular Weight (g/mol) 413.49
Formula Weight 413.49
Percent Purity 98.0%
Grade Research
Show More Show Less
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.