missing translation for 'onlineSavingsMsg'
Learn More

Obatoclax Mesylate, MedChemExpress

Product Code. 16404328
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16404328

Brand: Medchem Express HY10969

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Obatoclax Mesylate (GX15-070 Mesylate), a BH3 mimetic, is a pan-BCL-2 family proteins inhibitor with a Ki of 220 nM for BCL-2. Obatoclax Mesylate induces autophagy-dependent cell death and targets cyclin D1 for proteasomal degradation. Obatoclax Mesylate has anti-cancer and broad-spectrum antiparasitic activity.

Obatoclax Mesylate (GX15-070 Mesylate), a BH3 mimetic, is a pan-BCL-2 family proteins inhibitor with a Ki of 220 nM for BCL-2. Obatoclax Mesylate induces autophagy-dependent cell death and targets cyclin D1 for proteasomal degradation. Obatoclax Mesylate has anti-cancer and broad-spectrum antiparasitic activity.

Specifications

Chemical Name or Material Obatoclax Mesylate
Physical Form Solid
CAS 803712-79-0
Color Black
Quantity 10 mg
Purity Grade Notes Research
Molecular Formula C21H23N3O4S
For Use With (Application) Cancer-Kinase/protease
Synonym GX15-070 Mesylate
Solubility Information DMSO : 12.5 mg/mL (30.23 mM; Need ultrasonic)
SMILES COC1=CC(C2=CC3=C(C=CC=C3)N2)=N/C1=C\C4=C(C=C(N4)C)C.CS(=O)(O)=O
Molecular Weight (g/mol) 413.49
Formula Weight 413.49
Percent Purity 98.0%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.