missing translation for 'onlineSavingsMsg'
Learn More
Learn More
OAS2 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 19279-1-AP-150UL
This item is not returnable.
View return policy
Description
This antibody is specific to isoform p71 and isoform p69 of OAS2.
This gene encodes a member of the 2-5A synthetase family, essential proteins involved in the innate immune response to viral infection. The encoded protein is induced by interferons and uses adenosine triphosphate in 2′-specific nucleotidyl transfer reactions to synthesize 2′,5′-oligoadenylates. These molecules activate latent RNase L, which results in viral RNA degradation and the inhibition of viral replication. The three known members of this gene family are located in a cluster on chromosome 12. Alternatively spliced transcript variants encoding different isoforms have been described.Specifications
| OAS2 | |
| Polyclonal | |
| Unconjugated | |
| OAS2 | |
| (2 5)oligo(A) synthase 2, 2 5 oligoadenylate synthase 2, 2 5A synthase 2, OAS2, p69 OAS / p71 OAS, p69OAS / p71OAS | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 246728, 4939 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
| 0.33 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| E9Q9A9, P29728 | |
| OAS2 | |
| OAS2 Fusion Protein Ag6170 | |
| 150 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction