missing translation for 'onlineSavingsMsg'
Learn More
Learn More
OAS1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 14955-1-AP-150UL
Additional Details : Weight : 0.00010kg
Description
OAS1 is an interferon inducible protein that may play a role in mediating resistance to virus infection, control of cell growth, differentiation, and apoptosis. It binds double-stranded RNA and polymerizes ATP into PPP(A2′P5′A)N oligomers, which activate the latent RNase L that, when activated, cleaves single-stranded RNAs. This protein is associated with different subcellular fractions such as mitochondrial, nuclear, and rough/smooth microsomal fractions.Specifications
OAS1 | |
Polyclonal | |
Unconjugated | |
OAS1 | |
(2 5)oligo(A) synthase 1, 2 5 oligoadenylate synthase 1, 2 5A synthase 1, E18/E16, IFI 4, OAS1, OIAS, OIASI, p46/p42 OAS | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
192281, 246730, 4938 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
0.16 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P00973, P11928, Q05961 | |
OAS1, Oas1a | |
OAS1 Fusion Protein Ag6793 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |