missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ OAS1 Monoclonal Antibody (C5), Invitrogen™

Mouse Monoclonal Antibody
2060.00 NOK - 4530.00 NOK
Specifications
| Antigen | OAS1 |
|---|---|
| Clone | C5 |
| Concentration | 1 mg/mL |
| Applications | Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
17674096
|
Invitrogen™
MA543643 |
20 μL |
2060.00 NOK
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17664096
![]() |
Invitrogen™
MA543642 |
100 μL |
4530.00 NOK
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
OAS1 is an interferon inducible protein that may play a role in mediating resistance to virus infection, control of cell growth, differentiation, and apoptosis. It binds double-stranded RNA and polymerizes ATP into PPP(A2'P5'A)N oligomers, which activate the latent RNase L that, when activated, cleaves single-stranded RNAs. This protein is associated with different subcellular fractions such as mitochondrial, nuclear, and rough/smooth microsomal fractions.Specifications
| OAS1 | |
| 1 mg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4 | |
| (2-5')oligo(A) synthase 1; (2-5')oligo(A) synthase 1A; (2-5')oligo(A) synthetase 1; (2-5')oligo(A) synthetase 1A; 2',5'-oligo A synthetase 1; 2',5'-oligoadenylate synthetase 1, 40/46kDa; 25 oligoadenylate synthetase; 2'-5' oligoadenylate synthetase; 2'-5' oligoadenylate synthetase 1; 2'-5' oligoadenylate synthetase 1A; 2'-5' oligoadenylate synthetase 1G; 2-5A synthase 1; 2-5A synthase 1A; 2-5A synthetase 1; 2-5A synthetase 1A; 2'-5'-oligoadenylate synthase 1; 2'-5'-oligoadenylate synthase 1; LOW QUALITY PROTEIN: 2'-5'-oligoadenylate synthase 1; 2'-5'-oligoadenylate synthase 1A; 2'-5'-oligoadenylate synthetase 1; 2-5-oligoadenylate synthetase 1; 2'-5'-oligoadenylate synthetase 1, 40/46kDa; 2'-5'-oligoisoadenylate synthetase 1; E16 (2'-5') oligo A synthetase; E18/E16; IFI 4; IFI4; IFI-4; L3; LOW QUALITY PROTEIN: 2'-5'-oligoadenylate synthase 1; OAS 1; Oas1; Oas1a; Oas1g; OIAS; Oias1; OIASI; p42 OAS; p46/p42 OAS | |
| OAS1 | |
| IgG1 κ | |
| Protein A/G | |
| Antibody |
| C5 | |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| Unconjugated | |
| Mouse | |
| Human | |
| P00973 | |
| 4938 | |
| Ser74-Pro165 (Accession P00973), with N-terminal His Tag. | |
| Primary | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| OAS1 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title